Assay Details
| Gene Symbol: ocrl-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001128949.5 | NP_001122421.1 | 2 - 3 | 450 | 148 | |
| NM_001375161.2 | NP_001361846.1 | 10 - 11 | 1907 | 148 | ||
More Information
Set Membership: |
|

ocrl-1
Genomic Map