Assay Details
| Gene Symbol: eif-3.C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_060237.5 | NP_492638.1 | 1 - 2 | 102 | 76 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

eif-3.C| Gene Symbol: eif-3.C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_060237.5 | NP_492638.1 | 1 - 2 | 102 | 76 | |
Set Membership: |
|