Assay Details
| Gene Symbol: cfi-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_060243.4 | NP_492644.2 | 2 - 3 | 522 | 71 | |
| GenBank mRNA | AF487547.1 | - | 2 - 3 | 522 | 71 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

cfi-1| Gene Symbol: cfi-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_060243.4 | NP_492644.2 | 2 - 3 | 522 | 71 | |
| GenBank mRNA | AF487547.1 | - | 2 - 3 | 522 | 71 | |
Set Membership: |
|