Assay Details
| Gene Symbol: nubp-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_060252.7 | NP_492653.2 | 4 - 5 | 468 | 93 | |
| Gene Symbol: ercc-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001383709.1 | 4 - 5 | 470 | 93 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_060252.7 transcripts.
Set Membership: |
|

nubp-1
Genomic Map