Assay Details
| Gene Symbol: iars-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001264388.4 | NP_001251317.2 | 7 - 8 | 1211 | 95 | |
| Gene Symbol: C25A1.20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001383714.1 | 6 - 7 | 1261 | 95 | ||
More Information
Set Membership: |
|

iars-2
Genomic Map