Assay Details
| Gene Symbol: iars-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001264388.4 | NP_001251317.2 | 10 - 11 | 2717 | 94 | |
| Gene Symbol: C25A1.20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001383714.1 | 9 - 10 | 2764 | 94 | ||
More Information
Set Membership: |
|

iars-2
Genomic Map