Assay Details
| Gene Symbol: dao-5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025833.9 | NP_001021004.1 | 3 - 4 | 625 | 70 | |
| NM_001025834.6 | NP_001021005.1 | 3 - 4 | 609 | 70 | ||
More Information
Set Membership: |
|

dao-5
Genomic Map