Assay Details
| Gene Symbol: mus-101 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_060342.8 | NP_492743.3 | 2 - 3 | 185 | 107 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

mus-101| Gene Symbol: mus-101 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_060342.8 | NP_492743.3 | 2 - 3 | 185 | 107 | |
Set Membership: |
|