Assay Details
| Gene Symbol: F59C6.8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_060347.5 | NP_492748.3 | 7 - 8 | 1067 | 87 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

F59C6.8| Gene Symbol: F59C6.8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_060347.5 | NP_492748.3 | 7 - 8 | 1067 | 87 | |
Set Membership: |
|