Assay Details
| Gene Symbol: duox-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_058283.1 | NP_490684.1 | 15 - 16 | 3039 | 77 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

duox-2| Gene Symbol: duox-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_058283.1 | NP_490684.1 | 15 - 16 | 3039 | 77 | |
Set Membership: |
|