Assay Details
| Gene Symbol: ptr-11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001306284.3 | NP_001293213.1 | 6 - 7 | 1049 | 106 | |
| NM_058286.7 | NP_490687.1 | 11 - 12 | 1620 | 106 | ||
More Information
Set Membership: |
|

ptr-11
Genomic Map