Assay Details
| Gene Symbol: ptr-11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001306284.3 | NP_001293213.1 | 9 - 10 | 1861 | 76 | |
| NM_058286.7 | NP_490687.1 | 14 - 15 | 2432 | 76 | ||
More Information
Set Membership: |
|

ptr-11
Genomic Map