Assay Details
| Gene Symbol: ptr-11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001306284.3 | NP_001293213.1 | 2 - 3 | 149 | 132 | |
| NM_058286.7 | NP_490687.1 | 7 - 8 | 720 | 132 | ||
More Information
Set Membership: |
|

ptr-11
Genomic Map