Assay Details
| Gene Symbol: ptr-11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001306284.3 | NP_001293213.1 | 5 - 6 | 985 | 101 | |
| NM_058286.7 | NP_490687.1 | 10 - 11 | 1556 | 101 | ||
More Information
Set Membership: |
|

ptr-11
Genomic Map