Assay Details
| Gene Symbol: otub-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_058329.5 | NP_490730.2 | 4 - 5 | 489 | 105 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

otub-2| Gene Symbol: otub-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_058329.5 | NP_490730.2 | 4 - 5 | 489 | 105 | |
Set Membership: |
|