Assay Details
| Gene Symbol: otub-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001306333.3 | NP_001293262.1 | 3 - 4 | 493 | 83 | |
| NM_058329.5 | NP_490730.2 | 7 - 8 | 976 | 83 | ||
More Information
Set Membership: |
|

otub-2
Genomic Map