Assay Details
| Gene Symbol: ptr-21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_058350.5 | NP_490751.1 | 8 - 9 | 2506 | 86 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ptr-21| Gene Symbol: ptr-21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_058350.5 | NP_490751.1 | 8 - 9 | 2506 | 86 | |
Set Membership: |
|