Assay Details
| Gene Symbol: grl-16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_058353.5 | NP_490754.1 | 3 - 4 | 961 | 57 | |
| NM_182075.3 | NP_871875.1 | 3 - 4 | 1141 | 57 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

grl-16| Gene Symbol: grl-16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_058353.5 | NP_490754.1 | 3 - 4 | 961 | 57 | |
| NM_182075.3 | NP_871875.1 | 3 - 4 | 1141 | 57 | ||
Set Membership: |
|