Assay Details
| Gene Symbol: grl-16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_058353.5 | NP_490754.1 | 4 - 5 | 1066 | 66 | |
| NM_182075.3 | NP_871875.1 | 4 - 5 | 1246 | 66 | ||
More Information
Set Membership: |
|

grl-16
Genomic Map