Assay Details
| Gene Symbol: hasp-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_058367.5 | NP_490768.3 | 4 - 5 | 1203 | 72 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

hasp-2| Gene Symbol: hasp-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_058367.5 | NP_490768.3 | 4 - 5 | 1203 | 72 | |
Set Membership: |
|