Assay Details
| Gene Symbol: pif-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001306244.3 | NP_001293173.1 | 6 - 7 | 1077 | 67 | |
| NM_001306245.3 | NP_001293174.1 | 10 - 11 | 1608 | 67 | ||
| NM_001306246.3 | NP_001293175.1 | 1 - 2 | 135 | 67 | ||
| NM_058373.6 | NP_490774.2 | 12 - 13 | 2075 | 67 | ||
| GenBank mRNA | AB015041.1 | - | 12 - 13 | 1932 | 67 | |
More Information
Set Membership: |
|

pif-1
Genomic Map