Assay Details
| Gene Symbol: pde-6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001129083.4 | NP_001122555.1 | 3 - 4 | 498 | 71 | |
| NM_001383315.1 | 3 - 4 | 498 | 71 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

pde-6| Gene Symbol: pde-6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001129083.4 | NP_001122555.1 | 3 - 4 | 498 | 71 | |
| NM_001383315.1 | 3 - 4 | 498 | 71 | |||
Set Membership: |
|