Assay Details
| Gene Symbol: pde-6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001306261.3 | NP_001293190.1 | 3 - 4 | 1308 | 83 | |
| NM_001383315.1 | 9 - 10 | 2130 | 83 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

pde-6| Gene Symbol: pde-6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001306261.3 | NP_001293190.1 | 3 - 4 | 1308 | 83 | |
| NM_001383315.1 | 9 - 10 | 2130 | 83 | |||
Set Membership: |
|