Assay Details
| Gene Symbol: mig-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001026573.3 | NP_001021744.1 | 1 - 2 | 321 | 85 | |
| NM_058395.5 | NP_490796.2 | 1 - 2 | 307 | 85 | ||
More Information
Set Membership: |
|

mig-1
Genomic Map