Assay Details
| Gene Symbol: shc-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001383442.1 | 3 - 4 | 267 | 135 | ||
| NM_058398.5 | NP_490799.2 | 3 - 4 | 289 | 135 | ||
| NR_132552.1 | 1 - 2 | 68 | 135 | |||
| NR_132616.1 | 2 - 3 | 354 | 135 | |||
| Gene Symbol: cutl-20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_058397.7 | NP_490798.1 | 3 - 4 | 289 | 135 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001383442.1, NM_058397.7 transcripts.
Set Membership: |
|

shc-1
Genomic Map