Assay Details
| Gene Symbol: shc-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001383442.1 | 5 - 6 | 854 | 78 | ||
| NM_058398.5 | NP_490799.2 | 5 - 6 | 876 | 78 | ||
| NR_132552.1 | 3 - 4 | 655 | 78 | |||
| NR_132553.1 | 3 - 4 | 791 | 78 | |||
| NR_132554.1 | 5 - 6 | 4429 | 78 | |||
| NR_132615.1 | 5 - 6 | 931 | 78 | |||
| NR_132616.1 | 4 - 5 | 941 | 78 | |||
| Gene Symbol: cutl-20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_058397.7 | NP_490798.1 | 5 - 6 | 876 | 78 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_058397.7 transcripts.
Set Membership: |
|

shc-1
Genomic Map