Assay Details
Gene Symbol: smg-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_058428.6 | NP_490829.1 | 3 - 4 | 1085 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
