Assay Details
Gene Symbol: smg-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_058428.6 | NP_490829.1 | 6 - 7 | 2412 | 107 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
