Assay Details
| Gene Symbol: herc-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001306503.3 | NP_001293432.1 | 10 - 11 | 1577 | 108 | |
| NM_001306504.3 | NP_001293433.1 | 9 - 10 | 1442 | 108 | ||
| NM_001306505.3 | NP_001293434.1 | 7 - 8 | 1025 | 108 | ||
| NM_001306506.3 | NP_001293435.1 | 1 - 2 | 152 | 108 | ||
| NM_058433.5 | NP_490834.1 | 15 - 16 | 2580 | 108 | ||
More Information
Set Membership: |
|

herc-1
Genomic Map