Assay Details
| Gene Symbol: herc-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001306503.3 | NP_001293432.1 | 11 - 12 | 1766 | 98 | |
| NM_001306504.3 | NP_001293433.1 | 10 - 11 | 1631 | 98 | ||
| NM_001306505.3 | NP_001293434.1 | 8 - 9 | 1214 | 98 | ||
| NM_001306506.3 | NP_001293435.1 | 2 - 3 | 341 | 98 | ||
| NM_058433.5 | NP_490834.1 | 16 - 17 | 2769 | 98 | ||
More Information
Set Membership: |
|

herc-1
Genomic Map