Assay Details
| Gene Symbol: herc-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001306503.3 | NP_001293432.1 | 2 - 3 | 210 | 130 | |
| NM_001306504.3 | NP_001293433.1 | 1 - 2 | 75 | 130 | ||
| NM_058433.5 | NP_490834.1 | 7 - 8 | 1213 | 130 | ||
More Information
Set Membership: |
|

herc-1
Genomic Map