Assay Details
| Gene Symbol: grh-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001306482.4 | NP_001293411.1 | 1 - 2 | 565 | 74 | |
| NM_058436.5 | NP_490837.2 | 1 - 2 | 566 | 74 | ||
| GenBank mRNA | AY323527.1 | - | 1 - 2 | 565 | 74 | |
More Information
Set Membership: |
|

grh-1
Genomic Map