Assay Details
Gene Symbol: grh-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001306482.4 | NP_001293411.1 | 2 - 3 | 737 | 78 | |
NM_058436.5 | NP_490837.2 | 2 - 3 | 738 | 78 | ||
GenBank mRNA | AY323527.1 | - | 2 - 3 | 737 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
