Assay Details
| Gene Symbol: grh-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_058436.5 | NP_490837.2 | 4 - 5 | 1109 | 134 | |
| GenBank mRNA | AY323527.1 | - | 4 - 5 | 1108 | 134 | |
More Information
Set Membership: |
|

grh-1
Genomic Map