Assay Details
| Gene Symbol: dapk-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001306486.3 | NP_001293415.1 | 9 - 10 | 2994 | 63 | |
| NM_001306487.3 | NP_001293416.1 | 7 - 8 | 2289 | 63 | ||
| NM_058439.5 | NP_490840.2 | 11 - 12 | 3490 | 63 | ||
| GenBank mRNA | AY733040.1 | - | 11 - 12 | 3477 | 63 | |
More Information
Set Membership: |
|

dapk-1
Genomic Map