Assay Details
| Gene Symbol: dapk-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001306486.3 | NP_001293415.1 | 10 - 11 | 3537 | 71 | |
| NM_001306487.3 | NP_001293416.1 | 8 - 9 | 2832 | 71 | ||
| NM_001306488.3 | NP_001293417.1 | 6 - 7 | 2526 | 71 | ||
| NM_058439.5 | NP_490840.2 | 12 - 13 | 4033 | 71 | ||
| GenBank mRNA | AY733040.1 | - | 12 - 13 | 4020 | 71 | |
More Information
Set Membership: |
|

dapk-1
Genomic Map