Assay Details
| Gene Symbol: dapk-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001306486.3 | NP_001293415.1 | 5 - 6 | 1287 | 113 | |
| NM_001306487.3 | NP_001293416.1 | 3 - 4 | 582 | 113 | ||
| NM_001306488.3 | NP_001293417.1 | 2 - 3 | 708 | 113 | ||
| NM_058439.5 | NP_490840.2 | 7 - 8 | 1783 | 113 | ||
| GenBank mRNA | AY733040.1 | - | 7 - 8 | 1770 | 113 | |
More Information
Set Membership: |
|

dapk-1
Genomic Map