Assay Details
| Gene Symbol: bre-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001383737.1 | 6 - 7 | 1068 | 58 | ||
| NM_058471.2 | NP_490872.1 | 6 - 7 | 1073 | 58 | ||
| GenBank mRNA | AY130767.1 | - | 5 - 6 | 975 | 58 | |
| AY533306.1 | - | 5 - 6 | 975 | 58 | ||
More Information
Set Membership: |
|

bre-4
Genomic Map