Assay Details
| Gene Symbol: tln-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001262110.3 | NP_001249039.1 | 4 - 5 | 5143 | 75 | |
| NM_001313545.3 | NP_001300474.1 | 5 - 6 | 5969 | 75 | ||
| NM_001373653.2 | NP_001359949.1 | 9 - 10 | 7367 | 75 | ||
| NM_001383754.1 | 9 - 10 | 6682 | 75 | |||
| NM_058485.7 | NP_490886.2 | 8 - 9 | 7069 | 75 | ||
More Information
Set Membership: |
|

tln-1
Genomic Map