Assay Details
Gene Symbol: pcyt-2.1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001129065.2 | NP_001122537.1 | 7 - 8 | 837 | 65 | |
NM_001306413.3 | NP_001293342.1 | 6 - 7 | 662 | 65 | ||
NM_001306414.3 | NP_001293343.1 | 4 - 5 | 599 | 65 | ||
NM_001306415.3 | NP_001293344.1 | 2 - 3 | 284 | 65 | ||
NM_001306416.3 | NP_001293345.1 | 1 - 2 | 44 | 65 | ||
NM_001306419.2 | NP_001293348.1 | 8 - 9 | 875 | 65 | ||
NM_058530.5 | NP_490931.1 | 6 - 7 | 814 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
