Assay Details
| Gene Symbol: cdk-7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001383811.1 | 2 - 3 | 328 | 74 | ||
| GenBank mRNA | AF154004.1 | - | 2 - 3 | 259 | 74 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

cdk-7| Gene Symbol: cdk-7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001383811.1 | 2 - 3 | 328 | 74 | ||
| GenBank mRNA | AF154004.1 | - | 2 - 3 | 259 | 74 | |
Set Membership: |
|