Assay Details
| Gene Symbol: cdk-7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001383811.1 | 8 - 9 | 1024 | 119 | ||
| GenBank mRNA | AF154004.1 | - | 8 - 9 | 955 | 119 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

cdk-7| Gene Symbol: cdk-7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001383811.1 | 8 - 9 | 1024 | 119 | ||
| GenBank mRNA | AF154004.1 | - | 8 - 9 | 955 | 119 | |
Set Membership: |
|