Assay Details
| Gene Symbol: nsun-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_058557.4 | NP_490958.1 | 1 - 2 | 236 | 88 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

nsun-4| Gene Symbol: nsun-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_058557.4 | NP_490958.1 | 1 - 2 | 236 | 88 | |
Set Membership: |
|