Assay Details
| Gene Symbol: nsun-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001306435.3 | NP_001293364.1 | 2 - 3 | 642 | 73 | |
| NM_058557.4 | NP_490958.1 | 3 - 4 | 910 | 73 | ||
More Information
Set Membership: |
|

nsun-4
Genomic Map