Assay Details
| Gene Symbol: eif-2gamma | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_058559.8 | NP_490960.1 | 3 - 4 | 346 | 152 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

eif-2gamma| Gene Symbol: eif-2gamma | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_058559.8 | NP_490960.1 | 3 - 4 | 346 | 152 | |
Set Membership: |
|