Assay Details
| Gene Symbol: mcm-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_058561.6 | NP_490962.1 | 6 - 7 | 2175 | 72 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

mcm-4| Gene Symbol: mcm-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_058561.6 | NP_490962.1 | 6 - 7 | 2175 | 72 | |
Set Membership: |
|