Assay Details
| Gene Symbol: aat-5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_058601.6 | NP_491002.2 | 7 - 8 | 1196 | 88 | |
| NR_131358.1 | 7 - 8 | 1182 | 88 | |||
| NR_131359.1 | 6 - 7 | 1239 | 88 | |||
| NR_131780.1 | 7 - 8 | 1174 | 88 | |||
More Information
Set Membership: |
|

aat-5
Genomic Map