Assay Details
| Gene Symbol: ptr-12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001026388.5 | NP_001021559.2 | 8 - 9 | 1640 | 98 | |
| NR_131361.1 | 8 - 9 | 2193 | 98 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ptr-12| Gene Symbol: ptr-12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001026388.5 | NP_001021559.2 | 8 - 9 | 1640 | 98 | |
| NR_131361.1 | 8 - 9 | 2193 | 98 | |||
Set Membership: |
|