Assay Details
| Gene Symbol: spe-47 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001306358.3 | NP_001293287.1 | 1 - 2 | 76 | 110 | |
| NM_058609.3 | NP_491010.1 | 5 - 6 | 1067 | 110 | ||
More Information
Set Membership: |
|

spe-47
Genomic Map