Assay Details
| Gene Symbol: plk-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_058635.8 | NP_491036.1 | 3 - 4 | 713 | 77 | |
| GenBank mRNA | AF194964.1 | - | 3 - 4 | 735 | 77 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

plk-2| Gene Symbol: plk-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_058635.8 | NP_491036.1 | 3 - 4 | 713 | 77 | |
| GenBank mRNA | AF194964.1 | - | 3 - 4 | 735 | 77 | |
Set Membership: |
|